Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bub1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009773.3 | NP_033903.2 | 20 - 21 | 2876 | 79 | |
GenBank mRNA | AF107296.1 | - | 20 - 21 | 2831 | 79 | |
AK088502.1 | - | 6 - 7 | 932 | 79 | ||
AK133909.1 | - | 20 - 21 | 2817 | 79 | ||
BC031577.1 | - | 20 - 21 | 2827 | 4953223 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |