Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Dll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019454.3 | NP_062327.2 | 1 - 2 | 424 | 63 | |
GenBank mRNA | AB043893.1 | - | 1 - 2 | 67 | 63 | |
AF253469.1 | - | 1 - 2 | 379 | 63 | ||
AF273454.1 | - | 1 - 2 | 445 | 63 | ||
AK004739.1 | - | 1 - 2 | 392 | 63 | ||
AK031699.1 | - | 1 - 2 | 393 | 63 | ||
AK052322.1 | - | 1 - 2 | 425 | 63 | ||
AK165517.1 | - | 1 - 2 | 393 | 63 | ||
AK171717.1 | - | 1 - 2 | 393 | 63 | ||
BC042497.1 | - | 1 - 2 | 379 | 4017786 | 63 | |
BC049130.1 | - | 1 - 2 | 384 | 6825525 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |