Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029617.2 | NP_083893.2 | 6 - 7 | 373 | 121 | |
GenBank mRNA | AK009772.1 | - | 6 - 7 | 375 | 121 | |
AK033132.1 | - | 6 - 7 | 677 | 121 | ||
BC080815.1 | - | 6 - 7 | 691 | 30104270 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |