Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024145.2 | NP_001019316.1 | 4 - 5 | 552 | 88 | |
GenBank mRNA | AB195278.1 | - | 4 - 5 | 552 | 88 | |
AK076378.1 | - | 2 - 3 | 194 | 88 | ||
BC039947.1 | - | 4 - 5 | 401 | 88 | ||
BC145426.1 | - | 4 - 5 | 504 | 9053984 | 88 | |
BC145427.1 | - | 4 - 5 | 504 | 88 | ||
BC145428.1 | - | 4 - 5 | 507 | 9053986 | 88 | |
BC150771.1 | - | 4 - 5 | 507 | 9087682 | 88 | |
DQ904008.1 | - | 4 - 5 | 552 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |