Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200026316.mm.1 |
NM_001001714.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026316.mm.1 |
NM_001001714.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026316.mm.1 |
TC0200003169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026320.mm.1 |
NM_001001714.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026320.mm.1 |
NM_001001714.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026320.mm.1 |
TC0200003169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026317.mm.1 |
NM_001001714.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026317.mm.1 |
NM_001001714.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026317.mm.1 |
TC0200003169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026319.mm.1 |
NM_001001714.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026319.mm.1 |
NM_001001714.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026319.mm.1 |
TC0200003169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026318.mm.1 |
NM_001001714.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026318.mm.1 |
NM_001001714.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026318.mm.1 |
TC0200003169.mm.1 |
MTA TC |