Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sohlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001714.1 | NP_001001714.1 | 5 - 6 | 659 | 54 | |
GenBank mRNA | AY623913.1 | - | 5 - 6 | 659 | 54 | |
BC139189.1 | - | 5 - 6 | 627 | 8862211 | 54 | |
BC139190.1 | - | 5 - 6 | 627 | 8862212 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |