Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sohlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001714.1 | NP_001001714.1 | 6 - 7 | 847 | 89 | |
GenBank mRNA | AY623913.1 | - | 6 - 7 | 847 | 89 | |
BC139189.1 | - | 6 - 7 | 815 | 8862211 | 89 | |
BC139190.1 | - | 6 - 7 | 815 | 8862212 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |