Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290274.1 | NP_001277203.1 | 2 - 3 | 1325 | 70 | |
NM_153319.3 | NP_695231.3 | 2 - 3 | 1325 | 70 | ||
GenBank mRNA | AF461135.1 | - | 2 - 3 | 732 | 70 | |
BC057638.1 | - | 2 - 3 | 780 | 2631191 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |