Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022317.3 | NP_071712.3 | 4 - 5 | 444 | 70 | |
GenBank mRNA | AF305211.1 | - | 4 - 5 | 377 | 70 | |
AK052784.1 | - | 4 - 5 | 445 | 70 | ||
AK145077.1 | - | 4 - 5 | 445 | 70 | ||
AK164752.1 | - | 4 - 5 | 445 | 70 | ||
BC010472.1 | - | 4 - 5 | 432 | 3586005 | 70 | |
BC013783.1 | - | 4 - 5 | 435 | 3600786 | 70 | |
BY736278.1 | - | 4 - 5 | 445 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |