Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sppl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023220.2 | NP_075709.2 | 10 - 11 | 1331 | 96 | |
GenBank mRNA | AB041547.1 | - | 10 - 11 | 1331 | 96 | |
AK155011.1 | - | 10 - 11 | 1130 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mus musculus Probe spans exons |