Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277296.1 | NP_001264225.1 | 7 - 8 | 1135 | 92 | |
NM_001277297.1 | NP_001264226.1 | 7 - 8 | 947 | 92 | ||
NM_028105.4 | NP_082381.1 | 7 - 8 | 1056 | 92 | ||
GenBank mRNA | AK011310.1 | - | 7 - 8 | 907 | 92 | |
AK050868.1 | - | 7 - 8 | 1037 | 92 | ||
AK154839.1 | - | 7 - 8 | 1002 | 92 | ||
BC010539.1 | - | 7 - 8 | 949 | 4162365 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |