Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028875.2 | NP_083151.1 | 5 - 6 | 683 | 127 | |
GenBank mRNA | AK014491.1 | - | 5 - 6 | 683 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |