Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028875.2 | NP_083151.1 | 6 - 7 | 896 | 95 | |
GenBank mRNA | AK014491.1 | - | 6 - 7 | 896 | 95 | |
BC043073.1 | - | 5 - 6 | 773 | 6404521 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |