Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097621.1 | NP_001091090.1 | 8 - 9 | 1824 | 73 | |
GenBank mRNA | CB520233.1 | - | 3 - 4 | 586 | 73 | |
CD350871.1 | - | 2 - 3 | 332 | 73 | ||
GQ489027.1 | - | 8 - 9 | 1667 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |