Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097621.1 | NP_001091090.1 | 10 - 11 | 2169 | 76 | |
GenBank mRNA | AK129315.1 | - | 2 - 3 | 293 | 76 | |
CB518589.1 | - | 1 - 2 | 163 | 76 | ||
CD350871.1 | - | 4 - 5 | 677 | 76 | ||
GQ489027.1 | - | 10 - 11 | 2012 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |