Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291003.1 | NP_001277932.1 | 5 - 6 | 511 | 64 | |
NM_172539.3 | NP_766127.1 | 5 - 6 | 642 | 64 | ||
GenBank mRNA | AJ537599.2 | - | 5 - 6 | 450 | 64 | |
AK033037.1 | - | 5 - 6 | 642 | 64 | ||
BC064729.1 | - | 5 - 6 | 503 | 30476764 | 64 | |
DV649054.1 | - | 5 - 6 | 523 | 64 | ||
FJ187790.1 | - | 5 - 6 | 450 | 64 | ||
FJ187791.1 | - | 4 - 5 | 387 | 64 | ||
FJ187792.1 | - | 4 - 5 | 285 | 64 | ||
FJ187793.1 | - | 5 - 6 | 348 | 64 | ||
FJ187794.1 | - | 3 - 4 | 321 | 64 | ||
FJ187795.1 | - | 4 - 5 | 384 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |