Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB178898.1 | - | 1 - 2 | 85 | 154 | |
BC013531.1 | - | 1 - 2 | 135 | 4238756 | 154 | |
BC091761.1 | - | 1 - 2 | 135 | 5044038 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |