Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081364.3 | NP_001074833.3 | 3 - 4 | 599 | 90 | |
NM_001128084.2 | NP_001121556.2 | 3 - 4 | 599 | 90 | ||
GenBank mRNA | BC076629.1 | - | 3 - 4 | 599 | 30639338 | 90 |
BC150741.1 | - | 3 - 4 | 528 | 9087652 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |