Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177074.2 | NP_796048.2 | 9 - 10 | 959 | 74 | |
GenBank mRNA | AK043610.1 | - | 9 - 10 | 968 | 74 | |
AK139966.1 | - | 9 - 10 | 909 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons |