Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip4k2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008845.4 | NP_032871.3 | 1 - 2 | 265 | 63 | |
GenBank mRNA | AB009615.1 | - | 1 - 2 | 190 | 63 | |
AK012196.1 | - | 1 - 2 | 268 | 63 | ||
AK040558.1 | - | 1 - 2 | 155 | 63 | ||
BC011097.1 | - | 1 - 2 | 212 | 3672732 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |