Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015792.1 | NP_056607.1 | 16 - 17 | 2490 | 86 | |
GenBank mRNA | AF184275.1 | - | 14 - 15 | 1984 | 86 | |
AK083118.1 | - | 16 - 17 | 2835 | 86 | ||
AK084645.1 | - | 16 - 17 | 86 | |||
AK147829.1 | - | 16 - 17 | 2474 | 86 | ||
AK154357.1 | - | 16 - 17 | 2513 | 86 | ||
AK158937.1 | - | 16 - 17 | 2541 | 86 | ||
AK181430.1 | - | 2 - 3 | 121 | 86 | ||
BC004036.1 | - | 13 - 14 | 1576 | 86 | ||
BC031393.1 | - | 16 - 17 | 2490 | 4948804 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |