Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111289.1 | NP_001104759.1 | 15 - 16 | 2032 | 79 | |
NM_001111290.1 | NP_001104760.1 | 15 - 16 | 2005 | 79 | ||
NM_001111291.1 | NP_001104761.1 | 16 - 17 | 2072 | 79 | ||
NM_001111292.1 | NP_001104762.1 | 15 - 16 | 2032 | 79 | ||
NM_016739.3 | NP_058019.2 | 16 - 17 | 2072 | 79 | ||
GenBank mRNA | AB373955.1 | - | 15 - 16 | 1894 | 79 | |
AK011150.1 | - | 1 - 2 | 61 | 79 | ||
AK029283.1 | - | 1 - 2 | 63 | 79 | ||
AK031050.1 | - | 1 - 2 | 63 | 79 | ||
AK133891.1 | - | 16 - 17 | 2035 | 79 | ||
AK141992.1 | - | 15 - 16 | 1974 | 79 | ||
AK147514.1 | - | 13 - 14 | 6114 | 79 | ||
AK149183.1 | - | 2 - 3 | 273 | 79 | ||
AK157707.1 | - | 16 - 17 | 2056 | 79 | ||
AK161131.1 | - | 10 - 11 | 1167 | 79 | ||
AK166461.1 | - | 15 - 16 | 1990 | 79 | ||
AK168676.1 | - | 15 - 16 | 2000 | 79 | ||
AK169366.1 | - | 7 - 8 | 803 | 79 | ||
AK170830.1 | - | 16 - 17 | 2034 | 79 | ||
BC052427.1 | - | 16 - 17 | 2048 | 6836041 | 79 | |
BC127030.1 | - | 7 - 8 | 898 | 79 | ||
U18773.1 | - | 5 - 6 | 694 | 79 | ||
U27838.1 | - | 15 - 16 | 2009 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |