Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111289.1 | NP_001104759.1 | 2 - 3 | 411 | 156 | |
NM_001111290.1 | NP_001104760.1 | 2 - 3 | 411 | 156 | ||
NM_001111291.1 | NP_001104761.1 | 3 - 4 | 451 | 156 | ||
NM_001111292.1 | NP_001104762.1 | 2 - 3 | 411 | 156 | ||
NM_016739.3 | NP_058019.2 | 3 - 4 | 451 | 156 | ||
GenBank mRNA | AB373955.1 | - | 2 - 3 | 273 | 156 | |
AK133891.1 | - | 3 - 4 | 413 | 156 | ||
AK141992.1 | - | 2 - 3 | 353 | 156 | ||
AK147514.1 | - | 3 - 4 | 418 | 156 | ||
AK148203.1 | - | 2 - 3 | 398 | 156 | ||
AK157707.1 | - | 3 - 4 | 435 | 156 | ||
AK166461.1 | - | 2 - 3 | 369 | 156 | ||
AK168676.1 | - | 2 - 3 | 406 | 156 | ||
AK170830.1 | - | 3 - 4 | 413 | 156 | ||
BC052427.1 | - | 3 - 4 | 430 | 6836041 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |