Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111289.1 | NP_001104759.1 | 8 - 9 | 1089 | 68 | |
NM_001111290.1 | NP_001104760.1 | 8 - 9 | 1089 | 68 | ||
NM_001111291.1 | NP_001104761.1 | 9 - 10 | 1129 | 68 | ||
NM_001111292.1 | NP_001104762.1 | 8 - 9 | 1089 | 68 | ||
NM_016739.3 | NP_058019.2 | 9 - 10 | 1129 | 68 | ||
GenBank mRNA | AB373955.1 | - | 8 - 9 | 951 | 68 | |
AK133891.1 | - | 9 - 10 | 1092 | 68 | ||
AK141992.1 | - | 8 - 9 | 1031 | 68 | ||
AK147514.1 | - | 8 - 9 | 2900 | 68 | ||
AK148203.1 | - | 8 - 9 | 1076 | 68 | ||
AK157707.1 | - | 9 - 10 | 1113 | 68 | ||
AK161131.1 | - | 3 - 4 | 224 | 68 | ||
AK168676.1 | - | 8 - 9 | 1084 | 68 | ||
AK170830.1 | - | 9 - 10 | 1091 | 68 | ||
AK200310.1 | - | 2 - 3 | 119 | 68 | ||
BC052427.1 | - | 9 - 10 | 1108 | 6836041 | 68 | |
U27838.1 | - | 8 - 9 | 1066 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |