Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146047.2 | NP_666159.2 | 11 - 12 | 1435 | 104 | |
GenBank mRNA | AK032295.1 | - | 11 - 12 | 1399 | 104 | |
AK048357.1 | - | 11 - 12 | 1332 | 104 | ||
AK156203.1 | - | 11 - 12 | 1326 | 104 | ||
AK165939.1 | - | 11 - 12 | 1296 | 104 | ||
AK177383.1 | - | 6 - 7 | 426 | 104 | ||
AK181076.1 | - | 3 - 4 | 157 | 104 | ||
BC026562.1 | - | 11 - 12 | 1136 | 5028264 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |