Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190830.1 | NP_001177759.1 | 12 - 13 | 1894 | 81 | |
NM_010589.6 | NP_034719.2 | 12 - 13 | 1894 | 81 | ||
GenBank mRNA | AK038268.1 | - | 11 - 12 | 1705 | 81 | |
AK088365.1 | - | 12 - 13 | 1743 | 81 | ||
AK142178.1 | - | 12 - 13 | 1732 | 81 | ||
AK164520.1 | - | 12 - 13 | 1734 | 81 | ||
BC105577.1 | - | 11 - 12 | 1697 | 3968386 | 81 | |
BC131646.1 | - | 11 - 12 | 1705 | 40110146 | 81 | |
BC131647.1 | - | 11 - 12 | 1705 | 40110148 | 81 | |
L32955.1 | - | 11 - 12 | 1693 | 81 | ||
L33768.1 | - | 9 - 10 | 2335 | 81 | ||
L40172.1 | - | 12 - 13 | 1755 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |