Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011977.3 | NP_036107.1 | 3 - 4 | 374 | 75 | |
GenBank mRNA | AK040923.1 | - | 2 - 3 | 313 | 75 | |
AK138548.1 | - | 1 - 2 | 237 | 75 | ||
AK155185.1 | - | 1 - 2 | 180 | 75 | ||
AK164798.1 | - | 1 - 2 | 153 | 75 | ||
BC028937.1 | - | 3 - 4 | 374 | 5362790 | 75 | |
U15976.1 | - | 1 - 2 | 169 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |