Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029873.2 | NP_001025044.2 | 5 - 6 | 505 | 73 | |
GenBank mRNA | AK010728.1 | - | 4 - 5 | 441 | 73 | |
AK170263.1 | - | 4 - 5 | 1785 | 73 | ||
AY753536.1 | - | 5 - 6 | 395 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |