Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800005592.mm.1 |
NM_001145780.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005592.mm.1 |
NM_001145780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005592.mm.1 |
TC0800000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005598.mm.1 |
NM_001145780.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005598.mm.1 |
NM_001145780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005598.mm.1 |
NM_025917.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005598.mm.1 |
NM_025917.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005598.mm.1 |
TC0800000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005593.mm.1 |
NM_001145780.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005593.mm.1 |
NM_001145780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005593.mm.1 |
TC0800000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005602.mm.1 |
NM_001145780.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005602.mm.1 |
NM_001145780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005602.mm.1 |
NM_025917.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005602.mm.1 |
NM_025917.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005602.mm.1 |
TC0800000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005591.mm.1 |
NM_001145780.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005591.mm.1 |
NM_001145780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005591.mm.1 |
TC0800000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005603.mm.1 |
TC0800000783.mm.1 |
MTA TC |