Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmur2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153079.4 | NP_694719.2 | 2 - 3 | 893 | 71 | |
GenBank mRNA | AK036756.1 | - | 2 - 3 | 894 | 71 | |
AY057384.1 | - | 2 - 3 | 789 | 71 | ||
BC137611.1 | - | 2 - 3 | 822 | 8860630 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |