Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013463.2 | NP_038491.2 | 3 - 4 | 616 | 110 | |
GenBank mRNA | AK040081.1 | - | 3 - 4 | 548 | 110 | |
AK054547.1 | - | 3 - 4 | 568 | 110 | ||
AK145011.1 | - | 3 - 4 | 110 | |||
AK145148.1 | - | 110 | ||||
AK164231.1 | - | 3 - 4 | 549 | 110 | ||
AK166204.1 | - | 3 - 4 | 110 | |||
AK166245.1 | - | 3 - 4 | 110 | |||
BC009021.1 | - | 3 - 4 | 569 | 3708950 | 110 | |
U34071.1 | - | 3 - 4 | 566 | 110 | ||
U50716.1 | - | 3 - 4 | 554 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |