Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175446.3 | NP_780655.2 | 4 - 5 | 684 | 68 | |
GenBank mRNA | AK133258.1 | - | 636 | 68 | ||
AK220419.1 | - | 3 - 4 | 325 | 68 | ||
BC067022.1 | - | 4 - 5 | 660 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |