Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00007945.mm.1 |
NM_010078.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007945.mm.1 |
NM_010078.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007945.mm.1 |
TC0X00001277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007949.mm.1 |
NM_010078.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007949.mm.1 |
NM_010078.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007949.mm.1 |
TC0X00001277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007947.mm.1 |
NM_010078.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007947.mm.1 |
NM_010078.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007947.mm.1 |
TC0X00001277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007953.mm.1 |
NM_010078.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007953.mm.1 |
NM_010078.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007953.mm.1 |
TC0X00001277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007954.mm.1 |
NM_010078.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007954.mm.1 |
NM_010078.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007954.mm.1 |
TC0X00001277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007946.mm.1 |
NM_010078.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007946.mm.1 |
NM_010078.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007946.mm.1 |
TC0X00001277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007950.mm.1 |
NM_010078.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007950.mm.1 |
NM_010078.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007950.mm.1 |
TC0X00001277.mm.1 |
MTA TC |