Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC049191.1 | - | 10 - 11 | 1529 | 6476607 | 106 |
BC068151.1 | - | 9 - 10 | 1725 | 30534873 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |