Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012056.2 | NP_036186.2 | 3 - 4 | 711 | 64 | |
GenBank mRNA | AF090334.1 | - | 3 - 4 | 637 | 64 | |
AF279263.1 | - | 3 - 4 | 689 | 64 | ||
AK007499.1 | - | 1 - 2 | 101 | 64 | ||
BC026133.1 | - | 3 - 4 | 651 | 4018235 | 64 | |
BC043129.1 | - | 3 - 4 | 711 | 6810029 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |