Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030174.2 | NP_084450.2 | 4 - 5 | 628 | 95 | |
GenBank mRNA | AK047562.1 | - | 4 - 5 | 629 | 95 | |
AK154322.1 | - | 4 - 5 | 482 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |