Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021322.2 | NP_067297.2 | 9 - 10 | 1217 | 108 | |
GenBank mRNA | AJ271892.1 | - | 9 - 10 | 987 | 108 | |
AJ271893.1 | - | 9 - 10 | 987 | 108 | ||
AK035125.1 | - | 8 - 9 | 841 | 108 | ||
AK085245.1 | - | 4 - 5 | 431 | 108 | ||
AK144821.1 | - | 9 - 10 | 1022 | 108 | ||
AK152271.1 | - | 9 - 10 | 1009 | 108 | ||
AK153161.1 | - | 9 - 10 | 1009 | 108 | ||
AK164160.1 | - | 9 - 10 | 932 | 108 | ||
BC039272.1 | - | 9 - 10 | 1017 | 5027600 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |