Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC060663.1 | - | 9 - 10 | 1816 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |