Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027450.3 | NP_081726.1 | 3 - 4 | 279 | 81 | |
GenBank mRNA | AK017557.1 | - | 3 - 4 | 256 | 81 | |
AK154433.1 | - | 3 - 4 | 257 | 81 | ||
AK167595.1 | - | 3 - 4 | 279 | 81 | ||
BC031750.1 | - | 3 - 4 | 4037002 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |