Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024222.2 | NP_077184.2 | 3 - 4 | 1067 | 108 | |
GenBank mRNA | AK012153.1 | - | 3 - 4 | 736 | 108 | |
AK018758.2 | - | 3 - 4 | 777 | 108 | ||
AK145674.1 | - | 3 - 4 | 708 | 108 | ||
AK152899.1 | - | 3 - 4 | 1115 | 108 | ||
AK154979.1 | - | 3 - 4 | 756 | 108 | ||
AK170079.1 | - | 3 - 4 | 779 | 108 | ||
AX799084.1 | - | 3 - 4 | 778 | 108 | ||
BC052433.1 | - | 3 - 4 | 1067 | 6837097 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |