Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011758.2 | NP_035888.2 | 1 - 2 | 81 | ||
GenBank mRNA | AK045944.1 | - | 1 - 2 | 81 | ||
AK141135.1 | - | 1 - 2 | 81 | |||
BC108271.1 | - | 1 - 2 | 30102085 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |