Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276315.1 | NP_001263244.1 | 3 - 4 | 900 | 166 | |
NM_016912.2 | NP_058608.1 | 3 - 4 | 634 | 166 | ||
NM_177270.4 | NP_796244.2 | 3 - 4 | 634 | 166 | ||
GenBank mRNA | AB029065.1 | - | 3 - 4 | 610 | 166 | |
AB029066.1 | - | 3 - 4 | 598 | 166 | ||
AB029067.1 | - | 3 - 4 | 888 | 166 | ||
AK030598.1 | - | 3 - 4 | 889 | 166 | ||
AK139820.1 | - | 3 - 4 | 842 | 166 | ||
AK144574.1 | - | 3 - 4 | 635 | 166 | ||
AK145688.1 | - | 3 - 4 | 783 | 166 | ||
BC117894.1 | - | 3 - 4 | 543 | 40096852 | 166 | |
BC117895.1 | - | 3 - 4 | 543 | 40096853 | 166 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |