Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099631.1 | NP_001093101.1 | 7 - 8 | 953 | 64 | |
GenBank mRNA | AK032598.1 | - | 7 - 8 | 886 | 64 | |
BC036961.1 | - | 8 - 9 | 987 | 64 | ||
DQ832275.1 | - | 7 - 8 | 953 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |