Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Avl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030235.1 | NP_084511.1 | 1 - 2 | 309 | 63 | |
GenBank mRNA | AK020003.1 | - | 1 - 2 | 286 | 63 | |
AK083605.1 | - | 1 - 2 | 265 | 63 | ||
AK122228.1 | - | 1 - 2 | 287 | 63 | ||
AK147365.1 | - | 1 - 2 | 311 | 63 | ||
BC051177.1 | - | 1 - 2 | 291 | 63 | ||
BC117878.1 | - | 1 - 2 | 271 | 40095961 | 63 | |
BC117879.1 | - | 1 - 2 | 271 | 63 | ||
CJ109208.1 | - | 1 - 2 | 311 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |