Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177371.1 | NP_001170842.1 | 2 - 3 | 269 | 67 | |
NM_001177372.1 | NP_001170843.1 | 2 - 3 | 269 | 67 | ||
NM_019813.4 | NP_062787.2 | 2 - 3 | 269 | 67 | ||
GenBank mRNA | AB064321.1 | - | 2 - 3 | 259 | 67 | |
AF187147.1 | - | 2 - 3 | 259 | 67 | ||
AF187148.1 | - | 2 - 3 | 259 | 67 | ||
AK132060.1 | - | 2 - 3 | 255 | 67 | ||
AK162697.1 | - | 2 - 3 | 256 | 67 | ||
BC006714.1 | - | 3 - 4 | 217 | 3710497 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |