Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033245.4 | NP_001028417.1 | 9 - 10 | 1330 | 71 | |
NM_001206390.1 | NP_001193319.1 | 9 - 10 | 1140 | 71 | ||
NM_001206391.1 | NP_001193320.1 | 9 - 10 | 1140 | 71 | ||
NM_001206392.1 | NP_001193321.1 | 8 - 9 | 1210 | 71 | ||
GenBank mRNA | AK140444.1 | - | 9 - 10 | 1194 | 71 | |
AK149651.1 | - | 9 - 10 | 1180 | 71 | ||
AK149992.1 | - | 9 - 10 | 1142 | 71 | ||
AK152839.1 | - | 3 - 4 | 434 | 71 | ||
AK154603.1 | - | 9 - 10 | 1302 | 71 | ||
AK155701.1 | - | 9 - 10 | 1187 | 71 | ||
AK159857.1 | - | 8 - 9 | 1056 | 71 | ||
AK159906.1 | - | 9 - 10 | 1240 | 71 | ||
AK160018.1 | - | 9 - 10 | 1240 | 71 | ||
AK160063.1 | - | 9 - 10 | 1240 | 71 | ||
AK162637.1 | - | 9 - 10 | 1273 | 71 | ||
AK163658.1 | - | 9 - 10 | 1194 | 71 | ||
AK170813.1 | - | 8 - 9 | 1060 | 71 | ||
AK171214.1 | - | 9 - 10 | 1268 | 71 | ||
AK171248.1 | - | 9 - 10 | 1180 | 71 | ||
AK171580.1 | - | 9 - 10 | 1280 | 71 | ||
BC117860.1 | - | 9 - 10 | 1225 | 40095265 | 71 | |
BC117861.1 | - | 9 - 10 | 1225 | 40095267 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |