Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025992.2 | NP_080268.1 | 21 - 22 | 2728 | 113 | |
GenBank mRNA | AK147960.1 | - | 21 - 22 | 2651 | 113 | |
AK149648.1 | - | 21 - 22 | 2720 | 113 | ||
AK161293.1 | - | 9 - 10 | 1334 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |