Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178785.3 | NP_848900.2 | 10 - 11 | 1935 | 93 | |
GenBank mRNA | AK030797.1 | - | 10 - 11 | 1938 | 93 | |
AK040573.1 | - | 10 - 11 | 1923 | 93 | ||
AK088449.1 | - | 10 - 11 | 1897 | 93 | ||
AK088987.1 | - | 10 - 11 | 1913 | 93 | ||
AK131128.1 | - | 12 - 13 | 1842 | 93 | ||
BC132341.1 | - | 12 - 13 | 1841 | 40130618 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |