Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038018.4 | NP_001033107.1 | 3 - 4 | 442 | 67 | |
NM_001112711.2 | NP_001106182.1 | 3 - 4 | 442 | 67 | ||
NM_001286063.1 | NP_001272992.1 | 3 - 4 | 422 | 67 | ||
NM_001286064.1 | NP_001272993.1 | 3 - 4 | 442 | 67 | ||
NM_001286065.1 | NP_001272994.1 | 3 - 4 | 442 | 67 | ||
NM_001286066.1 | NP_001272995.1 | 3 - 4 | 442 | 67 | ||
NM_011938.4 | NP_036068.2 | 3 - 4 | 442 | 67 | ||
GenBank mRNA | AF040747.1 | - | 3 - 4 | 312 | 67 | |
AF040748.1 | - | 3 - 4 | 312 | 67 | ||
AF040749.1 | - | 3 - 4 | 312 | 67 | ||
AK051405.1 | - | 3 - 4 | 423 | 67 | ||
AK146119.1 | - | 3 - 4 | 317 | 67 | ||
AK161142.1 | - | 1 - 2 | 74 | 67 | ||
BC057206.1 | - | 3 - 4 | 431 | 67 | ||
BC075719.1 | - | 3 - 4 | 323 | 5031455 | 67 | |
Y15797.1 | - | 3 - 4 | 264 | 67 | ||
Y15798.1 | - | 3 - 4 | 286 | 67 | ||
Y15799.1 | - | 3 - 4 | 269 | 67 | ||
Y15800.1 | - | 3 - 4 | 264 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |