Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crebbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025432.1 | NP_001020603.1 | 25 - 26 | 4464 | 87 | |
GenBank mRNA | AK136057.1 | - | 20 - 21 | 2933 | 87 | |
BC072594.1 | - | 25 - 26 | 4450 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |